Learn More
TyK2-IN-2, MedChemExpress
Code produit 16496277
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16496277 5 mg
16489966 50 mg
16470899 1 mg
16464666 100 mg
16463558 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Code produit 16496277 Fournisseur Medchem Express Code fournisseur HY101762
MedChemExpress TyK2-IN-2 (Compoud 18) is a potent and selective TYK2 inhibitor with IC50s of 7 nM, 0.1 μM and 0.05 μM for TYK2 JH2, IL-23 and IFNα, respectively. TyK2-IN-2 also inhibits phosphodiesterase 4 (PDE4) with an IC50 of 62 nM. TyK2-IN-2 can be used for the research of inflammatory and autoimmune diseases.
TyK2-IN-2 (Compoud 18) is a potent and selective TYK2 inhibitor with IC50s of 7 nM, 0.1 μM and 0.05 μM for TYK2 JH2, IL-23 and IFNα, respectively. TyK2-IN-2 also inhibits phosphodiesterase 4 (PDE4) with an IC50 of 62 nM. TyK2-IN-2 can be used for the research of inflammatory and autoimmune diseases.
Spécification
Nom chimique ou matériau TyK2-IN-2
Forme physique Solid
CAS 2098466-94-3
Couleur Off-White
Quantité 5 mg
Notes de qualité de la pureté Research
Formule moléculaire C16H18N6O
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité DMSO : ≥ 25 mg/mL (80.55 mM)
SMILES CNC1=CC(NC2=CC(C)=CC(C)=C2)=NN3C1=NC=C3C(N)=O
Poids moléculaire (g/mol) 310.35
Poids de la formule 310.35
Pourcentage de pureté 98.0%
Qualité Research