Learn More
Unity Lab Services™ Verification Report Spectrophotometer
Product Code. 11861802
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
11861802 1 Day Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11861802 Supplier Unity Lab Services™ Supplier No. UMVP9313
 Restricted Item
Expédition estimée: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
  • Verification of wavelength accuracy using a Holmium filter (five peaks 279 to 637nm)
  • Verification of the accuracy and linearity of three-point absorbance at five wavelengths between 440nm and 635nm
  • Production of the verification report
  • Benefit using references D.K.D.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Spectrophotometers
Duration 1 Day
Product Type Verification Report
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.