missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Vertical Clean Bench

Product Code. 18861835 Shop All Labconco Corporation Products
Change view
Click to view available options
Tension:
230 V
Comprend:
UV Light, Protection Panel, Airflow Monitor
Certifications/Conformité:
CE
Unit Size:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tension Comprend Certifications/Conformité Unit Size
18861835 230 V UV Light, Protection Panel, Airflow Monitor CE 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18861835 Supplier Labconco™ Supplier No. 3660624
 Article restreint
Expédition estimée: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.

  • Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
  • Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
  • Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.

Specifications

Type de produit Vertical Clean Bench
Revêtement Powder
Type de filtre HEPA
Type d’éclairage UV
Matériau Powder-coated Aluminum and Steel
Certifications/Conformité CE
Couleur Glacier White, Gray
Profondeur (impériale) intérieure 25.3 in.
Profondeur (métrique) intérieure 64.3 cm
Description Vertical Laminar Flow
Hauteur (impériale) intérieure 30.2 in.
Hauteur (métrique) intérieure 81.3 cm
Comprend UV Light, Protection Panel, Airflow Monitor
Indice IP None
Tension 230 V
Largeur (impériale) 6 ft.
Largeur (impériale) intérieure 70.8 in.
Largeur (métrique) intérieure 179.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.