Learn More
Leica Microsystems Visoria B Laboratory Microscope for Hematopathology or Clinical Microbiology
Description
- Visoria B Laboratory Microscope (Global IVD) with illumination management
- Ergonomic tilting tube
- Two eyepieces (10x), field of view 22 mm
- High-power white LED
- Contrast methods: Brightfield, Darkfield, Phase
- Encoded nosepiece, 6x
- Objectives (N PLAN): 5x, 10x, 20x, 40x, 50x, 100x
- ErgoStage with Ceramic Plate, with specimen holder
- 3-step focus drive with height-adjustable focus knobs
- Condenser A 0.9 (P)
- Immersion liquid, ISO 8036
- Dust protection cover
- Quick start guide and user manual
- Efficiency through encoded functions
- More comfort with ergonomics
- Visualize sample details with the right contrast
Spécification
Spécification
| Eyepiece | 10X/20 mm |
| Type de tête | Trinocular, Tilting |
| Type d’éclairage | LED |
| Pouvoir grossissant | 5X to 40X |
| Modèle | Visoria B |
| Types d’objectifs | Plan |
| Type de produit | Binocular Microscope |