Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Universal
Code produit 30002154
Modifier l'affichage
Click to view available options
Quantité:
50 ml
Conditionnement:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30002154 50 ml 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30002154 Fournisseur R&D Systems™ Code fournisseur VCTS023
  • ONLINE EXCLUSIVE PRICE
    179.17€
    158.55€ / 50mL
    Économisez 20.62€
    11% de réduction
 Article restreint
Expédition estimée: 18-11-2026
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Store the unopened product at room temperature. Do not use past expiration date.
Gamme de produits R&D Systems
Quantité 50 ml