Learn More
W-54011, MedChemExpress
Code produit 16413267
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16413267 50 mg
16447248 10 mg
16441659 1 mg
16419777 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16413267 Fournisseur Medchem Express Code fournisseur HY16992A
MedChemExpress W-54011 is a potent and orally active non-peptide C5a receptor antagonist. W-54011 inhibits the binding of 125I-labeled C5a to human neutrophils with a Ki value of 2.2 nM. W-54011 also inhibits C5a-induced intracellular Ca2+ mobilization, chemotaxis, and generation of ROS in human neutrophils with IC50s of 3.1 nM, 2.7 nM, and 1.6 nM, respectively.
W-54011 is a potent and orally active non-peptide C5a receptor antagonist. W-54011 inhibits the binding of 125I-labeled C5a to human neutrophils with a Ki value of 2.2 nM. W-54011 also inhibits C5a-induced intracellular Ca2+ mobilization, chemotaxis, and generation of ROS in human neutrophils with IC50s of 3.1 nM, 2.7 nM, and 1.6 nM, respectively.
Spécification
Nom chimique ou matériau W-54011
CAS 405098-33-1
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C30H37ClN2O2
Quantité 50 mg
Informations sur la solubilité DMSO : ≥ 28 mg/mL (56.79 mM)
SMILES O=C(C1CCCC2=C1C=C(OC)C=C2)N(CC3=CC=C(N(C)C)C=C3)C4=CC=C(C(C)C)C=C4.[H]Cl
Poids moléculaire (g/mol) 493.08
Poids de la formule 493.08
Pourcentage de pureté 98.0%
Qualité Research
Forme physique Solid