Learn More
DWK LIFE SCIENCES™ Colonne de distillation de 200 mm Wheaton™
Code produit 12441561 Tous les produits DWK Life Sciences Produits
Modifier l'affichage
Click to view available options
Joint conique standard:
19/22
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Joint conique standard Conditionnement
12441561 19/22 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12441561 Fournisseur DWK LIFE SCIENCES™ Code fournisseur 385772
  • ONLINE EXCLUSIVE PRICE
    214.00€
    160.91€ / 1 pièces
    Économisez 53.09€
    24% de réduction
Expédition estimée: 25-02-2027
The WHEATON™ 200mm Distilling Column is designed for laboratory distillation applications, providing reliable separation and purification of liquids with a 200mm column length for consistent, reproducible results.
  • 200mm distilling column designed for laboratory distillation and liquid separation applications
  • Compatible with standard laboratory glassware setups for distillation procedures
  • Manufactured by DWK Life Sciences under the Wheaton brand, ensuring quality borosilicate glass construction
  • Suitable for purification and separation of liquid mixtures in research and analytical environments
Spécification
Type Colonne de distillation
Joint conique standard 19/22