Learn More
DiscoverX Wnt 3a protein
Code produit 17392429
Encompass
Change view
Click to view available options
:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit
17392429 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17392429 Fournisseur DiscoverX Code fournisseur 921100
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
WNT3A is a signaling glycoprotein that is important for multiple developmental processes, including cell fate regulation and patterning during embryogenesis, and cell-cell signaling during morphogenesis of developing neural tube. Also implicated in oncogenesis and adipogenesis.
WNT3A is a signaling glycoprotein that is important for multiple developmental processes, including cell fate regulation and patterning during embryogenesis, and cell-cell signaling during morphogenesis of developing neural tube. Also implicated in oncogenesis and adipogenesis. Binds receptors of the Frizzled family of transmembrane receptors.
Spécification
Quantité 10 μg
Type de produit Control Ligands