Learn More
Phenomenex™ Zebron Gas Management™ O-Rings
Code produit 31748605 Tous les produits Phenomenex Produits
Modifier l'affichage
Click to view available options
Conditionnement:
20 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Conditionnement
31748605 20 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 31748605 Fournisseur Phenomenex™ Code fournisseur AG63190
 Article restreint
Expédition estimée: 20-10-2026
Zebron Gas Management™ filters and traps remove contaminants from mobile phase gases which reduces the potential for column bleed, increases the lifetime of your GC column, and protects sensitive GC detectors from damage.
  • Brand: Zebron Gas Management
  • Product Type: O-Ring
TRUSTED_SUSTAINABILITY
Spécification
À utiliser avec (équipement) Click-On Inline Filter
Technique Gas Chromatography
Type O-rings