Learn More
Zika Virus (SPH2015) Envelope Antibody (35), Allophycocyanin, R&D Systems™
Product Code. 18610508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18610508 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18610508 Supplier Novus Biologicals Supplier No. NBP305856APC
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Zika Virus (SPH2015) Envelope Monoclonal specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Sandwich ELISA Capture.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Zika Virus (SPH2015) Envelope
Applications Sandwich ELISA Capture
Classification Monoclonal
Clone 35
Conjugate APC
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Product Type Antibody
Form Purified
Product Line Novus