CAS RN 1450806-98-0
CAS RN 1450806-98-0
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
résultats
1
MedChemExpress GLP-1(7-37) acetate
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.
2
MedChemExpress GLP-1(7-37) acetate
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.
3
MedChemExpress GLP-1(7-37) acetate
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.
4
MedChemExpress GLP-1(7-37) acetate
MedChemExpress GLP-1(7-37) acetate is an intestinal insulinotropic hormone that augments glucose induced insulin secretion.