CAS RN 17298-37-2
CAS RN 17298-37-2
IUPAC Nor:
3-(propanoyloxy)-4-(trimethylazaniumyl)butanoate
Synonymes:
(+-)-propionylcarnitine propionylcarnitine O-propionylcarnitine (r)-isomer propionylcarnitine (r)-propionylcarnitine propionylcarnitine, (R)- propionylcarnitine, (r)-isomer propionylcarnitine, (r)- (R)-propionylcarnitine propionyl carnitine
Poids moléculaire (g/mol):
217.27
Formule moléculaire:
C10H19NO4
InChi Key:
UFAHZIUFPNSHSL-UHFFFAOYNA-N
SMILES:
CCC(=O)OC(CC([O-])=O)C[N+](C)(C)C
1
–
1
de
1
résultats
1
Propionylcarnitine, MedChemExpress
MedChemExpress Propionylcarnitine is metabolized by carnitine acetyltransferase from propionyl-CoA. Increased propionylcarnitine is regarded as a biomarker of vitamin B12 deficiency.