CAS RN 190383-13-2
CAS RN 190383-13-2
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress Protease-Activated Receptor-2, amide
MedChemExpress Protease-Activated Receptor-2, amide (SLIGKV-NH2) is a highly potent protease-activated receptor-2 (PAR2) activating peptide.
2
MedChemExpress Protease-Activated Receptor-2, amide
MedChemExpress Protease-Activated Receptor-2, amide (SLIGKV-NH2) is a highly potent protease-activated receptor-2 (PAR2) activating peptide.
3
MedChemExpress Protease-Activated Receptor-2, amide
MedChemExpress Protease-Activated Receptor-2, amide (SLIGKV-NH2) is a highly potent protease-activated receptor-2 (PAR2) activating peptide.