CAS RN 218916-52-0
CAS RN 218916-52-0
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol
MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol is one of the lathyrane diterpenoids, that has anti-cancer activity.
2
MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol
MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol is one of the lathyrane diterpenoids, that has anti-cancer activity.