CAS RN 548-19-6
CAS RN 548-19-6
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
Isoginkgetin, MedChemExpress
MedChemExpress Isoginkgetin is a pre-mRNA splicing inhibitor inhibitor. Isoginkgetin also inhibits activities of both Akt, NF-κB and MMP-9. Isoginkgetin inhibits the activity of the 20S proteasome, induces apoptosis and activates autophagy.
2
Tocris Bioscience™ Isoginkgetin
Pre-mRNA splicing inhibitor; cell permeable