CAS RN 62054-67-5
CAS RN 62054-67-5
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
THS-044, MedChemExpress
MedChemExpress THS-044 binding stabilizes the HIF2(alpha) PAS-B folded state, for regulating HIF2 activity in endogenous and clinical settings.