CAS RN 6606-65-1
CAS RN 6606-65-1
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
Enbucrilate, Technical Grade, TRC
CAS: 6606-65-1 Formule moléculaire: C8H11NO2 Poids moléculaire (g/mol): 153.18 Synonyme: 2-Cyano-2-propenoic Acid Butyl Ester,2-Cyano-acrylic Acid Butyl Ester,BK 200,BK 201,BK 201 (adhesive),BK 300,BK 301,BK 301 (adhesive),Butyl 2-cyanoacrylate,Butyl cyanoacrylate,Butyl α-cyanoacrylate,Enbucrilate,Glubranh 1,Histacryl Blue,Hystoacryl,Indermil,Indermil Tissue Adhesive,InteguSeal Clear,Integuseal,Integuseal Microbial Sealant,Sicomet 6000,Tisuacryl,n-Butyl 2-cyanoacrylate Nom IUPAC: butyl 2-cyanoprop-2-enoate SMILES: COC(=O)C1C(c2ccccc2)(O1)c3ccccc3
2
Enbucrilate, MedChemExpress
MedChemExpress Enbucrilate (Butyl cyanoacrylate) is a cyanoacrylate ester that has been used as surgical tissue adhesive.