Learn More
2-Phenylpropane-d12, CDN
Click to view available options
Quantité:
0.25 g
0.5 g
Conditionnement:
0.25g
0.50g
Specifications
Specifications
| Nom chimique ou matériau | Isopropylbenzene-D12 |
| CAS | 97732-82-6 |
| Formule moléculaire | C9 D12 |
| Quantité | 0.5 g |
| Synonyme | Benzene-1,2,3,4,5-d5, 6-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (ACI), 6-[1-(Methyl-d3)ethyl-1,2,2,2-d4]benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, [1-(methyl-d3)ethyl-1,2,2,2-d4]- (9 CI), Cumene-d12, Perdeuterated dimethylphenylbenzene, [1-(Methyl-d3)ethyl-1,2,2,2-d4]benzene-d5, Isopropylbenzene-d12, Isopropylbenzene-D12, 2-Phenylpropane-d12 |
| Formule InChI | InChI=1 S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| Nom IUPAC | 1,2,3,4,5-pentadeuterio-6-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)benzene |
| Poids moléculaire (g/mol) | 132.2655 |
| Poids de la formule | 132.1692 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction