Learn More
Tocris Bioscience™ Temsirolimus
Click to view available options
:
10mg
Description
Temsirolimus is a mTOR inhibitor; inhibits tumor growth in breast cancer cell lines (IC50 values are 1.6 and 4.3 nM for SKBr3 and BT474, respectively). Inhibits HIF-1α-mediated VEGF production in breast cancer cell lines (BT474 and MDA-MB-231). Directly inhibits serum and VEGF mediated endothelial cell proliferation and morphogenesis in vitro and vessel formation in vivo. Causes G1/S cell cycle arrest in multiple cancer cell lines. Antiangiogenic. Activates autophagy.
Spécification
Spécification
| Quantité | 10 mg |
| Type de produit | Temsirolimus |
| Pureté | >98% |
| Poids moléculaire | 1030.29 |
| Inhibiteurs | Temsirolimus |
Usage exclusivement réservé à la recherche