missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse NGFR/TNFRSF16 (Q9Z0W1) VersaClone cDNA
Click to view available options
Quantité:
10 μg
Conditionnement:
10µg
Description
The Mouse NGF R/TNFRSF16 (Q9Z0W1) VersaClone cDNA clone contains the complete ORF for NGFR/TNFRSF16/p75NTR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The NGFR/TNFRSF16/p75NTR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spécification
Spécification
| Type de produit | cDNA |
| Expression | mNgfr |
| Numéro d’adhésion | Q9Z0W1 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Type d’insert | 1267bp |
| Identification génétique (Entrez) | 4804 (Human); 18053 (Mouse); 117089 (Rat) |
| Alias de gène | CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Mouse |
| Afficher plus |
Usage exclusivement réservé à la recherche
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu