missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 110 kDa FITC-dextran Green Fluorophore
Click to view available options
Quantité:
100 mg
1 g
5 g
Conditionnement:
100mg
1g
5g
Description
- Product Overview: FITC-dextrans are dextran molecules covalently labeled with fluorescein isothiocyanate (FITC), providing bright green fluorescence ideal for permeability studies, molecular tracking, and bioimaging applications. Available in a wide range of molecular weights from 4 kDa up to 2000 kDa, FITC-dextrans are widely used in studies of barrier integrity, vascular leakage, macromolecular diffusion, and cellular uptake in biological systems.
- Features:
- Bright Green Fluorescence: Excitation at 495 nm and emission at 519 nm, compatible with standard FITC filter sets for microscopy, plate readers, and flow cytometry.
- Wide Molecular Weight Range: Available from 4 kDa to 2000 kDa to suit studies involving different diffusion rates and molecular size exclusion properties.
- Water-Soluble and Biocompatible: Fully soluble in aqueous solutions, enabling direct use in cell culture, tissue studies, and in vivo models.
- High Lot-to-Lot Consistency: Manufactured with controlled degree of labeling and tight molecular weight distribution for reproducibility across experiments.
- Flexible Detection Methods: Suitable for both live imaging and endpoint assays involving fluorescence microscopy, spectrophotometry, or flow cytometry.
- Benefits:
- Quantitative Analysis of Barrier Function: Widely used to measure permeability across cellular monolayers, tissues, and engineered barriers.
- Modeling Molecular Diffusion: Different molecular weights allow precise modeling of size-dependent diffusion across biological membranes and matrices.
- Tracking Macromolecular Transport: Useful for studying vascular permeability, blood-brain barrier integrity, and nanoparticle delivery models.
- Suitable for Live and Fixed Systems: Can be applied in live cell experiments as well as fixed tissue studies without significant loss of fluorescence.
- Standardized Fluorescence Signal: FITC labeling ensures a strong, predictable fluorescence signal across different imaging platforms.
- Applications:
- Barrier Permeability Assays: Used to evaluate tight junction function and paracellular transport in epithelial and endothelial models.
- Vascular Leakage Studies: Applied in models of inflammation, stroke, cancer, and injury to assess changes in vascular permeability.
- Macromolecular Diffusion Research: Traces molecular movement in microfluidic devices, 3D cultures, and organoids.
- Cellular Uptake and Endocytosis Studies: Used to monitor macropinocytosis, vesicular trafficking, and intracellular transport dynamics.
- Preclinical Drug Delivery Studies: Models the behavior of therapeutic macromolecules in biological systems.
- Technical Specifications:
- Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm.
- Molecular Weights: Available at 4 kDa, 10 kDa, 20 kDa, 40 kDa, 70 kDa, 110 kDa, 150 kDa, 500 kDa, and 2000 kDa options.
- Appearance: Supplied as a light-sensitive, greenish powder, readily soluble in water.
- Degree of Labeling: Optimized for bright signal without altering dextran diffusion behavior significantly.
- Storage and Stability:
- Store dry, protected from light, at 2°C to 8°C for maximum stability.
- Stable under recommended conditions for extended storage periods without significant degradation of fluorescence intensity.
- Packaging and Availability:
- Supplied as dry powder for easy handling and storage.
- Available in a range of packaging sizes to accommodate small-scale experiments and large research projects.
Spécification
Spécification
| Quantité | 5 g |
| Type de produit | Fluorescent Labeling Reagent |
| Poids moléculaire | 110 kDa |
| Type d’étiquette | Other Labels or Dyes |
| Couleur | Green |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu