Learn More
Chem-Lab 1LT Potassium pyroantimonate solution 13.33 g KSb(OH)6 + 16.65 g KOH + 0.57 g NaOH / l H2O
Marque:  Chem-Lab CL07.1103.1000
1LT Potassium pyroantimonate solution 13.33 g KSb(OH)6 + 16.65 g KOH + 0.57 g NaOH / l H2O
Code produit. 18833674
  • 85.50€ / 1L