Learn More
Carbon Steel, AISI 1020, G10200, ARMI∣MBH
Code produit. 17908834
Click to view available options
Conditionnement:
1 pièce
Code produit. 17908834
Marque: ARMI/MBH IARMFE102018G
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Carbon Steel, AISI 1020, G10200, Ø38x19mm
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Niobium/Antimony/Calcium/Aluminium/Tantalum/Zinc/Arsenic/Phosphorus/Cobalt/Oxygen/Tin/Nitrogen/Sulfur/Molybdenum/Vanadium/Nickel/Chromium/Copper/Carbon/Silicon/Manganese/Iron/Cadmium/Bismuth/Magnesium/Selenium/Hydrogen/Lead/Boron/Tungsten/Titanium/Zirconium
CAS 7439-89-6
Forme Disc
Concentration or Composition (by Analyte or Components) Niobium 0.0012 %/Antimony 0.0018 %/Calcium 0.002 %/Aluminium 0.003 %/Tantalum 0.003 %/Zinc 0.003 %/Arsenic 0.0044 %/Phosphorus 0.006 %/Cobalt 0.0065 %/Oxygen 0.007 %/Tin 0.008 %/Nitrogen 0.0098 %/Sulfur 0.024 %/Molybdenum 0.0252 %/Vanadium 0.036 %/Nickel 0.078 %/Chromium 0.125 %/Copper 0.198 %/Carbon 0.226 %/Silicon 0.235 %/Manganese 0.547 %/Iron 98.5 %/Cadmium (<0.0010) %/Bismuth (<0.0050) %/Magnesium (<0.0050) %/Selenium (<0.0050) %/Hydrogen (0.0002) %/Lead (0.0003) %/Boron (0.0006) %/Tungsten (0.0007) %/Titanium (0.001) %/Zirconium (0.002) %