Learn More
3-Hydroxy-3-methylpentane-1,5-dioic Acid, TRC
Code produit 30133301
Change view
Click to view available options
Quantité:
100 mg
500 mg
1 g
Conditionnement:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30133301 1 g 1g
30137750 500 mg 500mg
30004815 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30133301 Fournisseur Toronto Research Chemicals Code fournisseur TRCH9482631G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 503-49-1
Formule moléculaire C6 H10 O5
Poids moléculaire (g/mol) 162.14
Synonyme Pentanedioic acid, 3-hydroxy-3-methyl-, Glutaric acid, 3-hydroxy-3-methyl- (6CI,7CI,8CI), 3-Hydroxy-3-methylpentanedioic acid, 3-Hydroxy-3-methylglutaric acid, 3-Methyl-3-hydroxyglutaric acid, 3-Methyl-3-hydroxypentanedioic acid, CB 337, Dicrotalic acid, HMG, HMGA, Lipoglutaren, Medroglutaric acid, Meglutol, Mevalon, NSC 361411, β-Hydroxy-β-methylglutaric acid
Nom IUPAC 3-hydroxy-3-methylpentanedioic acid
SMILES CC(O)(CC(=O)O)CC(=O)O
Specifications
Nom chimique ou matériau 3-Hydroxy-3-methylpentane-1,5-dioic Acid
Notes de qualité de la pureté HPLC
Quantité 1 g
Formule InChI InChI=1S/C6H10O5/c1-6(11,2-4(7)8)3-5(9)10/h11H,2-3H2,1H3,(H,7,8)(H,9,10)
Poids de la formule 162.0528
Pourcentage de pureté >95%