missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-biphényl)-2-(4-phényle-2-pyridyl)thiazole, 97 %, Thermo Scientific™
337.00€ - 1374.00€
Identifiants chimiques
CAS | 1858241-90-3 |
---|---|
Formule moléculaire | C26H18N2S |
Poids moléculaire (g/mol) | 390.504 |
Numéro MDL | MFCD22682975 |
Clé InChI | VMTLWMDKPVRQEX-UHFFFAOYSA-N |
Synonyme | 4-4-biphenylyl-2-4-phenyl-2-pyridyl thiazole, 2-4-1,1'-biphenyl-4-yl-1,3-thiazol-2-yl-4-phenylpyridine |
CID PubChem | 86277493 |
Nom IUPAC | 4-(4-phénylphényl)-2-(4-phénylpyridine-2-yl)-1,3-thiazole |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)C4=NC=CC(=C4)C5=CC=CC=C5 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
1858241-90-3 | |
390.504 | |
VMTLWMDKPVRQEX-UHFFFAOYSA-N | |
86277493 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)C4=NC=CC(=C4)C5=CC=CC=C5 |
C26H18N2S | |
MFCD22682975 | |
4-4-biphenylyl-2-4-phenyl-2-pyridyl thiazole, 2-4-1,1'-biphenyl-4-yl-1,3-thiazol-2-yl-4-phenylpyridine | |
4-(4-phénylphényl)-2-(4-phénylpyridine-2-yl)-1,3-thiazole |
Spécification
4-(4-Biphenylyl)-2-(4-phenyl-2-pyridyl)thiazole | |
C26H18N2S | |
4-4-biphenylyl-2-4-phenyl-2-pyridyl thiazole, 2-4-1,1'-biphenyl-4-yl-1,3-thiazol-2-yl-4-phenylpyridine | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)C4=NC=CC(=C4)C5=CC=CC=C5 | |
390.504 | |
390.5 |
1858241-90-3 | |
MFCD22682975 | |
VMTLWMDKPVRQEX-UHFFFAOYSA-N | |
4-(4-phénylphényl)-2-(4-phénylpyridine-2-yl)-1,3-thiazole | |
86277493 | |
97% |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures