missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ester pinacolique de l’acide 4-Fluoro-2-méthylbenzèneboronique, 96 %, Thermo Scientific™
41.10€ - 144.00€
Identifiants chimiques
CAS | 815631-56-2 |
---|---|
Formule moléculaire | C13H18BFO2 |
Poids moléculaire (g/mol) | 236.09 |
Numéro MDL | MFCD05863911 |
Clé InChI | HHWOQSZBVGPYMH-UHFFFAOYSA-N |
Synonyme | 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid pinacol ester, 4-fluoro-2-methylphenylboronic acid, pinacol ester, 2-4-fluoro-2-methylphenyl 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid,pinacol ester, 4-fluoro-2-methylbenzeneboronic acid pinacol ester, 2-4-fluoranyl-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl |
CID PubChem | 2759037 |
Nom IUPAC | 2-(4-fluoro-2-méthylphényl)-4,4,5,5-tétraméthyle-1,3,2-dioxaborolane |
SMILES | CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Price | Quantity & Availability | |||||
15457056
|
Thermo Scientific Alfa Aesar
H62120.03 |
1 g |
41.10€
1g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
15467056
|
Thermo Scientific Alfa Aesar
H62120.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
815631-56-2 | |
236.09 | |
HHWOQSZBVGPYMH-UHFFFAOYSA-N | |
2759037 | |
CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1 |
C13H18BFO2 | |
MFCD05863911 | |
2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid pinacol ester, 4-fluoro-2-methylphenylboronic acid, pinacol ester, 2-4-fluoro-2-methylphenyl 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid,pinacol ester, 4-fluoro-2-methylbenzeneboronic acid pinacol ester, 2-4-fluoranyl-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl | |
2-(4-fluoro-2-méthylphényl)-4,4,5,5-tétraméthyle-1,3,2-dioxaborolane |
Specifications
4-Fluoro-2-methylbenzeneboronic acid pinacol ester | |
C13H18BFO2 | |
2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid pinacol ester, 4-fluoro-2-methylphenylboronic acid, pinacol ester, 2-4-fluoro-2-methylphenyl 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-methylphenylboronic acid,pinacol ester, 4-fluoro-2-methylbenzeneboronic acid pinacol ester, 2-4-fluoranyl-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-fluoro-2-methyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-4-fluoro-2-methylphenyl-4,4,5,5-tetramethyl | |
CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1 | |
236.09 | |
236.09 | |
Solide |
815631-56-2 | |
MFCD05863911 | |
HHWOQSZBVGPYMH-UHFFFAOYSA-N | |
2-(4-fluoro-2-méthylphényl)-4,4,5,5-tétraméthyle-1,3,2-dioxaborolane | |
2759037 | |
96% |
Safety and Handling
missing translation for 'tsca' : Non
Recommended Storage : Température ambiante