Learn More
Tocris Bioscience™ 5-Ph-IAA-AM
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
5-Ph-IAA-AM is a potent and selective "Bump & Hole" TAG Degrader, for use within an auxin-inducible degron (AID2) system; it is an egg-permeant analog of 5-Ph-IAA (Cat. No. 7392). 5-Ph-IAA-AM induces selective degradation of a target protein tagged with a single-point mutant AtTIR1F79G F-box protein. 5-Ph-IAA-AM induces target protein degradation in C. elegans embryos. No developmental abnormality is observed when the embryos are treated with 50 μM 5-Ph-IAA-AM.
Spécification
Spécification
| Nom chimique ou matériau | Acetoxymethyl 2-(5-phenyl-1H-indol-3-yl)acetate |
| Quantité | 10 mg |
| Cible | TAG Degradation Platforms |
| Formule moléculaire | C19H17NO4 |
| Pureté | 0.98 |