missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetamide, TRC
Specifications
Specifications
| Nom chimique ou matériau | Acetoacetamide |
| Noms d’analytes ou de composants | Acetoacetamide |
| Notes de qualité de la pureté | HPLC |
| Conditions d’expédition | Room Temperature |
| Formule InChI | InChI=1S/C4H7NO2/c1-3(6)2-4(5)7/h2H2,1H3,(H2,5,7) |
| Type de solution | Neat |
| Industry Type | Forensics |
| Quantité | 10 g |
| Poids de la formule | 101.0477 |
| Pourcentage de pureté | >95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?