missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACO2 (Human) Recombinant Protein
Human ACO2 full-length ORF ( NP_001089.1, 1 a.a. - 780 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000050-P01.10ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Sequence: MAPYSLLVTRLQKALGVRQYHVASVLCQRAKVAMSHFEPNEYIHYDLLEKNINIVRKRLNRPLTLSEKIVYGHLDDPASQEIERGKSYLRLRPDRVAMQDATAQMAMLQFISSGLSKVAVPSTIHCDHLIEAQVGGEKDLRRAKDINQEVYNFLATAGAKYGVGFWKPGSGIIHQIILENYAYPGVLLIGTDSHTPNGGGLGGICIGVGGADAVDVMAGIPWELKCPKVIGVKLTGSLSGWSSPKDVILKVAGILTVKGGTGAIVEYHGPGVDSISCTGMATICNMGAEIGATTSVFPYNHRMKKYLSKTGREDIANLADEFKDHLVPDPGCHYDQLIEINLSELKPHINGPFTPDLAHPVAEVGKVAEKEGWPLDIRVGLIGSCTNSSYEDMGRSAAVAKQALAHGLKCKSQFTITPGSEQIRATIERDGYAQILRDLGGIVLANACGPCIGQWDRKDIKKGEKNTIVTSYNRNFTGRNDANPETHAFVTSPEIVTALAIAGTLKFNPETDYLTGTDGKKFRLEAPDADELPKGEFDPGQDTYQHPPKDSSGQHVDVSPTSQRLQLLEPFDKWDGKDLEDLQILIKVKGKCTTDHISAAGPWLKFRGHLDNISNNLLIGAINIENGKANSVRNAVTQEFGPVPDTARYYKKHGIRWVVIGDENYGEGSSREHAALEPRHLGGRAIITKSFARIHETNLKKQGLLPLTFADPADYNKIHPVDKLTIQGLKDFTPGKPLKCIIKHPNGTQETILLNHTFNETQIEWFRAGSALNRMKELQQ
Spécification
NP_001089.1 | |
aconitase 2, mitochondrial | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACO2 | |
GST |
50 | |
Wheat germ expression system | |
10 μg | |
ACONM, MGC20605, MGC33908 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |