Learn More
Invitrogen™ Human ACVR1 (ALK2), GST Tag Recombinant Protein
Code produit. p-7121652
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
Code produit. 15408693
Marque: Invitrogen™ PR8866A
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY
Spécification
Numéro d’adhésion Q04771
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50 mM tris with 0.02% polysorbate 20, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Identification génétique (Entrez) 90
Poids moléculaire 69.1 kDa
Nom Human ACVR1 (ALK2), GST Tag
Quantité 100 μg
État réglementaire RUO - research use only
Alias de gène activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7 L; TSRI; TSR-I; type I TGF B receptor
Nom usuel ACVR1
Famille des kinases STKR Kinase Family
Longueur des protéines 145-509
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Système d’expression Insect cells
Forme Liquid