missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADCK3, GST Tag Recombinant Protein

Code produit. p-7181238
Click to view available options
Quantité:
1 mg
10 μg
100 μg
Conditionnement:
100µg
10µg
1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15980293

Marque: Invitrogen™ A30969

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ADCK3 recombinant human protein (156-end) was expressed in insect cells. The purified catalytic domain (135-end) of ADCK3 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. ADCK3 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Catalytic.

ADCK3 serine/threonine kinase is a mitochondrial protein similar to yeast ABC1 that is a member of the ADCK family. CABC1 encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q8NI60
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris HCl with 25% glycerol, 10mM glutathione, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA, 50mM NaCl and no preservative; pH 7.5
Identification génétique (Entrez) 56997
Poids moléculaire 82.3 kDa
Nom Human ADCK3, GST Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 10 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Alias de gène 4632432J16Rik; aarF domain containing kinase 3; aarF domain-containing protein kinase 3; Adck3; AI462003; ARCA2; atypical kinase ADCK3, mitochondrial; Atypical kinase COQ8A, mitochondrial; Cabc1; Chaperone activity of bc1 complex-like; chaperone activity of bc1 complex-like, mitochondrial; chaperone, ABC1 activity of bc1 complex homolog; chaperone, ABC1 activity of bc1 complex like; Chaperone-ABC1-like; Coenzyme Q protein 8A; coenzyme Q8 homolog; coenzyme Q8A; COQ10D4; COQ8; COQ8A; mKIAA0451; PP265; SCAR9
Nom usuel CABC1
Symbole de gène(s) COQ8A
Famille des kinases ACK Kinase Family
Longueur des protéines 135-end
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag
Séquence 135-end
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Conditions d’expédition Dry Ice
Système d’expression Insect cells
Famille de protéines Kinases & Inhibitors
Forme protéique Recombinant, Catalytic
Forme Liquid
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis