Learn More
DiscoverX ADP QuestTM
Click to view available options
Quantité:
8 ml (800 dp, 384 puits)
Conditionnement:
1 kit
Description
The ADP QuestTM kit is a detection kit for the measurement of ADP produced as a product of kinase activity. It is used to detect substrate-induced activity of purified or semi-purified kinases. ADP produced by the kinase reacts with intermediate factors to produce a fluorescent dye precursor, which generates signal when combined with peroxide and peroxidase. This detection kit contains reagents for 2 plates (384-well).
Spécification
Spécification
| Entrée du dosage | Kinase activity |
| À utiliser avec (application) | Detection Kits |
| Quantité | 8 ml (800 dp, 384 puits) |
| Entrée cible | Any kinase, ATPase, GTPase or UTPase |
| Cible | Any kinase, ATPase, GTPase or UTPase |