Learn More
InSphero Akura™ PLUS Hanging Drop System
Code produit. 17921002
Click to view available options
Nbre de puits:
96
Conditionnement:
10 pièces
Code produit. 17921002
Marque: InSphero CSPF24
Il en reste null
Ajouter au panier
Ajouter au panier
The world-famous and unique Akura™ PLUS Hanging Drop System in a 96-well format with automation-compatible top-loading.
The world-famous and unique Akura™ PLUS Hanging Drop System in a 96-well format with automation-compatible top-loading.
  • Loading of cell suspension from the top through unique SureDrop™ inlet engineered for highly uniform and stable hanging drops.
  • Gravity-assisted formation of uniform spheroid/organoid morphology and functionality in long-term cell culture results in one spheroid per well.
  • Suitable for many cell types, including cells that don't normally form spheroids or organoids on ultra-low attachment surfaces.
  • Contains 10 Akura™ PLUS Hanging Drop Plates and 10 Akura™ 96 Spheroid Plates, individually wrapped, made of polystyrene and cyclo olefin polymer.
Spécification
Matériau Polystyrène
Volume (métrique) du puits 40 μL
Volume (métrique) utile 40 μL
Type Microplaque
Nbre de puits 96
Jetable Single-use
Autoclavable Not Autoclavable
Exempt de RNase Yes