Learn More
Abnova™ ALDH6A1 (Human) Recombinant Protein
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
- Sequence: VRARILQVSSKVKSSPTWYSASSFSSSVPTVKLFIGGKFVESKSDKWIDIHNPATNEVIGRVPQATKAEMDAAIASCKRAFPAWADTSVLSRQQVLLRYQQLIKENLKEIAKLITLEQGKTLADAEGDVFRGLQVVEHACSVTSLMMGETMPSITKDMDLYSYRLPLGVCAGIAPFNFPAMIPLWMFPMAMVCGNTFLMKPSERVPGATMLLAKLLQDSGAPDGTLNIIHGQHEAVNFICDHPDIKAISFVGSNKAGEYIFERGSRHGKRVQANMGAKNHGVVMPDANKENTLNQLVGAAFGAAGQRCMALSTAVLVGEAKKWLPELVEHAKNLRVNAGDQPGADLGPLITPQAKERVCNLIDSGTKEGASILLDGRKIKVKGYENGNFVGPTIISNVKPNMTCYKEEIFGPVLVVLETETLDEAIQIVNNNPYGNGTAIFTTNGATARKYAHLVDVGQVGVNVPIPVPLPMFSFTGSRSSFRGDTNFYGKQGIQFYTQLKTITSQWKEEDATLSSPAVVMPTMGR
Spécification
Spécification
| Numéro d’adhésion | AAH32371.1 |
| Identification génétique (Entrez) | 4329 |
| Nom | aldehyde dehydrogenase 6 family, member A1 |
| Méthode de préparation | Wheat germ expression system |
| Test du contrôle qualité | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantité | 10 μg |
| Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Alias de gène | MGC40271, MMSADHA, MMSDH |
| Symbole de gène(s) | ALDH6A1 |
| Espèces | Wheat Germ (in vitro) |
| Afficher plus |