missing translation for 'onlineSavingsMsg'
Learn More

Aliskiren, MedChemExpress

Code produit. p-200102033
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16451247

Marque: Medchem Express HY12176

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress Aliskiren is an orally active, highly potent and selective renin inhibitor, with IC50 of 1.5 nM. Aliskiren can be used for the research of hypertension, cardiovascular diseases and cancer cachexia.

Aliskiren is an orally active, highly potent and selective renin inhibitor, with IC50 of 1.5 nM. Aliskiren can be used for the research of hypertension, cardiovascular diseases and cancer cachexia.

Specifications

Nom chimique ou matériau Aliskiren
CAS 173334-57-1
Couleur Incolore
Notes de qualité de la pureté Research
Formule moléculaire C30H53N3O6
Quantité 50 mg
Synonyme CGP 60536 CGP60536B SPP 100
Informations sur la solubilité Ethanol : 100 mg/mL (181.24 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (181.24 mM; Need ultrasonic)
SMILES COCCCOC1=CC(C[C@@H](C[C@@H]([C@H](C[C@H](C(NCC(C)(C(N)=O)C)=O)C(C)C)O)N)C(C)C)=CC=C1OC
Poids moléculaire (g/mol) 551.76
Poids de la formule 551.76
Pourcentage de pureté 95.24%
Qualité Research
Forme physique Oil
À utiliser avec (application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.