missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amsacrine, MedChemExpress
Click to view available options
Quantité:
10 mg
50 mg
100 mg
500 mg
Description
Amsacrine (m-AMSA
Spécification
Spécification
| Nom chimique ou matériau | Amsacrine |
| CAS | 51264-14-3 |
| Couleur | Red |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C21H19N3O3S |
| Quantité | 100 mg |
| Synonyme | m-AMSA acridinyl anisidide |
| Informations sur la solubilité | DMSO : 9.3 mg/mL (23.64 mM; Need ultrasonic and warming) |
| SMILES | CS(=O)(NC1=CC=C(NC2=C(C=CC=C3)C3=NC4=CC=CC=C42)C(OC)=C1)=O |
| Poids moléculaire (g/mol) | 393.46 |
| Afficher plus |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction