Learn More
Tocris Bioscience™ Amyloid beta-Peptide (1-40) (human)
Code produit. 18739696
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
Code produit. 18739696
Marque: Tocris Bioscience™ 1191/1
Amyloid β-protein fragment
Amyloid β-Peptide (1-40) (human) is a peptide found in plaques in the brains of patients with Alzheimer's disease. Shown to have both neurotrophic and neurotoxic effects.
TRUSTED_SUSTAINABILITY
Spécification
Description Amyloid b-Peptide (1-40) (human)
Poids moléculaire 4329.86
Type de produit Amyloid β-Peptide (1-40) (human)
Quantité 1 mg
Séquence Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val
Usage exclusivement réservé à la recherche