Learn More
Tocris Bioscience™ Apcin
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
Apcin is a Cdc20 inhibitor; inhibits Cdc20-substrate interaction. Blocks substrate-mediated Cdc20 loading onto the anaphase-promoting complex (APC). Inhibits APC-dependent ubiquitylation. Prolongs mitotic duration in RPE1 cells in combination with proTAME (prodrug of TAME (Cat.No. 4506)) in vitro.
Specifications
Specifications
| Nom chimique ou matériau | 3-(2-Methyl-5-nitroimidazol-1-yl)-N-(2,2,2-trichloro-1-phenylaminoethyl)propionamide |
| CAS | 300815-04-7 |
| Quantité | 10 mg |
| Cible | Ubiquitin Ligase (E3) Inhibitors |
| Formule moléculaire | C13H14Cl3N7O4 |
| Poids de la formule | 438.65 |
| Pureté | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction