Learn More
CPC (Colder) AseptiQuik™ S Low-Flow Genderless Quick-Disconnect Fittings, Sanitary Adapter, High-Temperature Polycarbonate
Click to view available options
Comprend:
0,25 po Sanitary Clamp Size
0,75 po Taille de la pince sanitaire
Conditionnement:
1 pièce
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Specifically designed for small-volume fluid transfer applications. Robust construction provides reliable and repeatable fluid transfer. Sterilize by autoclave up to 130°C (266°F). 0.25 in. nominal flow path size
Spécification
Spécification
| Certifications/Conformité | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Température min. de fonctionnement (métrique) | 1°C |
| Quantité d’emballage | 1 de chaque |
| Port de sortie | Quick Disconnect x Sanitary |
| Méthode de stérilisation | Gamma irradiation up to 50 kGy |
| Forme | Structuré |
| Comprend | 0,25 po Sanitary Clamp Size |
| Matériau (joint) | Platinum-cured Silicone |
| Couleur | White with blue pull tab |