Learn More
Astemizole, TRC
Code produit 17941515
Change view
Click to view available options
Quantité:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
17941515 50 mg 50mg
17920935 25 mg 25mg
17934865 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941515 Supplier Toronto Research Chemicals Supplier No. TRCA79003050MG
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68844-77-9
Formule moléculaire C28 H31 F N4 O
Poids moléculaire (g/mol) 458.57
Synonyme 1H-Benzimidazol-2-amine, 1-[(4-fluorophenyl)methyl]-N-[1-[2-(4-methoxyphenyl)ethyl]-4-piperidinyl]-, Astemisan, Astemizole, Hismanal, Histamen, Histaminos, Histazol, Kelp, Laridal, Metodik, NSC 329963, Novo-Nastizol A, Paralergin, R 42512, R 43512, Retolen, Waruzol
Nom IUPAC 1-[(4-fluorophenyl)methyl]-N-[1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl]benzimidazol-2-amine
SMILES COc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1
Specifications
Nom chimique ou matériau Astemizole
Noms d’analytes ou de composants Astemizole
Notes de qualité de la pureté HPLC
Conditions d’expédition Room Temperature
Formule InChI InChI=1S/C28H31FN4O/c1-34-25-12-8-21(9-13-25)14-17-32-18-15-24(16-19-32)30-28-31-26-4-2-3-5-27(26)33(28)20-22-6-10-23(29)11-7-22/h2-13,24H,14-20H2,1H3,(H,30,31)
Type de solution Neat
Industry Type Forensics
Quantité 50 mg
Poids de la formule 458.2482
Pourcentage de pureté >95%