Learn More
Agilent BioTek AutoScratch Wound Making Tool
Tous les produits Agilent BioTek Produits
Click to view available options
À utiliser avec (équipement):
Lionheart FX, Cytation 5, Cytation 1
Conditionnement:
1 pièce
Description
- Automatically creates reproducible scratch wounds in cell monolayers grown in microplates.
- The simple push button operation and tool-free scratch pin manifold exchange make it easy to process either 96 or 24 well plates which are commonly used in migration and invasion assays.
- The compact system features an onboard pre-programmed cleaning routine to keep the scratch pins free of buildup and avoiding contamination.
- AutoScratch precisely and efficiently automates the sample prep for imaging workflows with Agilent BioTeks Cytation Cell Imaging Multi-Mode Readers and Lionheart Automated Microscopes.
- For complete capture through data publication workflows pre-defined protocols in Gen5 software automatically calculate wound width wound % confluence and maximum wound healing rate.
Spécification
Spécification
| À utiliser avec (application) | Wound Healing and Cell Migration Assays |
| À utiliser avec (équipement) | Lionheart FX, Cytation 5, Cytation 1 |