Learn More
Tocris Bioscience™ AZ 10397767
missing translation for 'orderingAttributeHoverText'
Quantité:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
AZ 10397767 is a potent CXCR2 antagonist (IC50 = 1 nM); attenuates oxaliplatin -induced NF-κB activation, increases oxaliplatin cytotoxicity, and potentiates oxaliplatin-induced apoptosis in AIPC cells. Reduces the numbers of neutrophils infiltrating into tumors in both in vitro and in vivo models and delayed tumor growth. Orally bioavailable.
Specifications
Specifications
| CAS | 333742-63-5 |
| Quantité | 10 mg |
| Cible | Chemokine Receptor |
| Formule moléculaire | C15H14ClFN4O2S2 |
| Poids de la formule | 400.88g/mol |
| Type de produit | Antagonist |
Usage exclusivement réservé à la recherche
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction