Learn More
Invitrogen™ Réactif d’histone H3 BacMam.
Code produit. 12073569
Click to view available options
Quantité:
100 ml
Conditionnement:
100mL
Code produit. 12073569
Marque: Invitrogen™ A14154
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
The BacMam Histone H3 Reagent provides a convenient genetic delivery tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest.
The BacMam Histone H3 Reagent provides a convenient genetic delivery tool for the expression of GFP-Histone H3 fusion protein in your cell background of interest. The GFP fusion enables sensitive detection of post-translational modifications using LanthaScreen™ TR-FRET Cellular Assay technology.
The BacMam Histone H3 Reagent Lets You:
  • Use your cell background of choice with the portability of BacMam
  • Conserve precious cell samples with the miniaturizable homogenous assay format
  • Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
  • Improve data quality with the advantages of TR-FRET
Get More Physiologically Relevant Results
The BacMam Histone H3 allows the investigation of post-translational modifications of histone H3 in your choice of cellular backgrounds, including primary cells. This enables screening for potential inhibitors of histone-modifying enzymes such as kinases, acetylases, deacetylases, methyl transferases, and demethylases in their natural complexes in a physiologically relevant cell type.
BacMam and LanthaScreen™ Convenience Saves Sample and Time
When used with a LanthaScreen™ Tb-anti-Histone H3 antibody against a specific modification, assays can be run in a fully homogenous, addition-only format without any of the washing, lysate transfer, or separation procedures required for traditional methods such as Western blotting and ELISA. By using the same BacMam reagent for GFP-Histone H3, a variety of histone H3 modifications (Ph, Me, Ac) can be monitored just by exchanging the Terbium (Tb) labeled antibody. In addition, the application of the LanthaScreen™ technology includes all of the advantages of TR-FRET detection, including reduced data noise, less interference from fluorescent compounds, and high sensitivity, allowing the use of fewer cells.
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Conserver le réactif BacMam histone H3 à 4°C
Type de livraison Baculovirus (système BacMam)
Quantité 100 ml
Type d’échantillon Liquide
Conditions d’expédition Glace humide