missing translation for 'onlineSavingsMsg'
Learn More

Barnidipine hydrochloride, MedChemExpress

Code produit. 16474108
Encompass_Preferred
Click to view available options
Quantité:
10 mg
50 mg
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16474108

Marque: Medchem Express HY107322

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors. Barnidipine hydrochloride (Mepirodipine hydrochloride) is an antihypertensive drug and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action.

Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors.

Specifications

Nom chimique ou matériau Barnidipine hydrochloride
CAS 104757-53-1
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C27H30ClN3O6
Quantité 10 mg
Synonyme Mepirodipine hydrochloride YM-09730-5
Informations sur la solubilité DMSO : 250 mg/mL (473.48 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)[C@@H]1C2=CC([N+]([O-])=O)=CC=C2)O[C@@H]3CN(CC4=CC=CC=C4)CC3.[H]Cl
Poids moléculaire (g/mol) 528.0
Poids de la formule 528.0
Pourcentage de pureté 98.77%
Qualité Research
Forme physique Solid
À utiliser avec (application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.