Learn More
BD Human Cell Surface Marker Screening Panel
Code produit 15840459
Modifier l'affichage
Click to view available options
Quantité:
5 Tests
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
15840459 5 Tests 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 15840459 Fournisseur BD Code fournisseur 560747
 Article restreint
Expédition estimée: 02-10-2026
Includes
Three 96 well plates, each well containing 2.75μg of antibody, enough for five tests (0.5μg/test) along with AlexaFluor™ 647 conjugated goat anti-mouse Ig and goat anti-rat Ig secondary antibodies
For screening cell lines, primary cells or tissue, and is compatible with flow cytometry and bioimaging technology platforms
Bioimaging, Flow Cytometry
TRUSTED_SUSTAINABILITY
Spécification
Quantité 5 Tests