Description
The Begain Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spécification
Spécification
| Lignée cellulaire | The lysate was created in HEK293T cells, using plasmid ID RC2943 and based on accession number NM_2836. The protein contains a C-terminal DDK tag. |
| À utiliser avec (application) | Western Blot |
| Quantité | 0.1 mg |
| Tampon de stockage | RIPA buffer |
| Pureté | Protein |
| Spécificité du test | Homo sapiens brain-enriched guanylate kinase-associated homolog (rat) (BEGAIN), mRNA. |
| Contenu et stockage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Usage exclusivement réservé à la recherche