Learn More
Tocris Bioscience™ Biotin-LC-NAD+
Biotinylated analog of β-NAD+; commonly used reagent in LSD1 and PARP1 assays
Marque:  Tocris Bioscience™ 6572/118U
Code produit. 18737826
  • 357.44€ / 118µg
Découvrez plus d'offres spéciales
Description
Description
Biotin-LC-NAD+ is a biotinylated analog of β-NAD+. Commonly used reagent in LSD1 and PARP1 assays.
Spécification
Spécification
β-Nicotinamide-N6-(2-(6-(6-[biotinyl]aminohexanoyl)aminohexanoyl)aminoethyl)adenine dinucleotide sodium salt
118 μg
C45H67N12NaO18P2S
95%