Learn More
ACROBiosystems Biotinylated Recombinant Protein L Protein, His Tag, ultra sensitivity (primary amine labeling, long spacer)
Recombinant Protein
Spécification
Source | Biotinylated Recombinant Protein L, His Tag, primary amine labeling, long spacer (RPL-P814R) is expressed from E.coli cells. It is the biotinylated form of Recombinant Protein L, His Tag (RPL-P3141). |
---|---|
Conditions de stockage | -20°C |
Alias de gène | RPL,Protein L |
Nom usuel | Protein L |
Conjugué | UltraLys |
Description
Protein L was isolated from the surface of bacterial species Peptostreptococcus magnus and was found to bind Ig(IgG,IgM,IgA,IgE and IgD) through L chain interaction, from which the name was suggested. Despite this wide-ranging binding capability with respect to Ig classes, Protein L is not a universal immunoglobilin-binding protein. Binding of Protein L to immunoglobulins is restricted to those containing kappa light chains (i.e., k chain of the VL domain). In humans and mice, kappa (k) light chains predominate. The remaining immunoglobulins have lambda (l) light chains. The recombinant protein contains four immunoglobulin (Ig) binding domains (Bdomains) of the native protein. Besides antibody, protein L is also suitable for binding of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies (Dabs).
This protein carries a polyhistidine tag at the N-terminus. The protein migrates as 50-60 kDa on a SDS-PAGE gel under reducing (R) condition. This protein can bind to VL-Kappa.Spécification
Biotinylated Recombinant Protein L, His Tag, primary amine labeling, long spacer (RPL-P814R) is expressed from E.coli cells. It is the biotinylated form of Recombinant Protein L, His Tag (RPL-P3141). | |
RPL,Protein L | |
UltraLys | |
PBS, pH7.4 | |
E.coli | |
MABSol™ Biotin Labeled Proteins |
-20°C | |
Protein L | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |