missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BV6, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.
Specifications
Specifications
| Nom chimique ou matériau | BV6 |
| CAS | 1001600-56-1 |
| Couleur | Blanc |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C70H96N10O8 |
| Quantité | 5 mg |
| Informations sur la solubilité | DMSO : ≥ 58 mg/mL (48.11 mM) |
| SMILES | O=C(NCCCCCCNC([C@@H](NC([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)=O)C(C3=CC=CC=C3)C4=CC=CC=C4)=O)[C@@H](NC([C@H](CCC5)N5C([C@H](C6CCCCC6)NC([C@H](C)NC)=O)=O)=O)C(C7=CC=CC=C7)C8=CC=CC=C8 |
| Poids moléculaire (g/mol) | 1205.57 |
| Poids de la formule | 1205.57 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction