Learn More
Tocris Bioscience™ CBA
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
CBA is a selective TRPM4 blocker (IC50 = 1.5 μM). Exhibits no significant activity against TRPM5, TRPM7, TRPM8, TRPV1, TRPV3, TRPV6 and a range of other ion channels and receptors. Displays neuroprotective effects against glutamate-induced neurodegeneration in vitro.
Spécification
Spécification
| Nom chimique ou matériau | 4-Chloro-2-[[2-(2-chlorophenoxy)acetyl]amino]benzoic acid |
| CAS | 351424-20-9 |
| Quantité | 10 mg |
| Formule moléculaire | C15H11Cl2NO4 |
| Poids de la formule | 340.16 |
| Pureté | 98% |